missing translation for 'onlineSavingsMsg'
Learn More

Vinyl Propionate (stabilized with MEHQ) 98.0+%, TCI America™

Catalog No. P0613500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0613500ML 500 mL
P061325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0613500ML Supplier TCI America Supplier No. P0613500ML

Chemical Identifiers

CAS 105-38-4
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.117
MDL Number MFCD00027004
InChI Key UIWXSTHGICQLQT-UHFFFAOYSA-N
Synonym vinyl propionate, propanoic acid, ethenyl ester, vinyl propanoate, propionic acid vinyl ester, propionic acid, vinyl ester, vinylpropionate, vinylester kyseliny propionove, vinylester kyseliny propionove czech, propanoic acid, ethenyl ester, homopolymer, propionic acid vinyl
PubChem CID 7750
IUPAC Name ethenyl propanoate
SMILES CCC(=O)OC=C

Specifications

Melting Point -80°C
Color Colorless
Boiling Point 95°C
Quantity 500 mL
UN Number 3272
Formula Weight 100.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Vinyl Propionate (stabilized with MEHQ)
EINECSNumber (2)-0758
RTECSNumber UF8575000
TSCA Yes