missing translation for 'onlineSavingsMsg'
Learn More

Vinyl Hexanoate (stabilized with MEHQ) 99.0+%, TCI America™

Catalog No. p-7124673
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C089825ML 25 mL
C0898500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C089825ML Supplier TCI America Supplier No. C089825ML

Chemical Identifiers

CAS 3050-69-9
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.20
MDL Number MFCD00048869
InChI Key LZWYWAIOTBEZFN-UHFFFAOYSA-N
Synonym Hexanoic Acid Vinyl Ester
PubChem CID 76451
IUPAC Name ethenyl hexanoate
SMILES CCCCCC(=O)OC=C

Specifications

Color Colorless
Boiling Point 167°C
Quantity 25 mL
UN Number 3272
Formula Weight 142.20
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Vinyl Hexanoate (stabilized with MEHQ)
EINECSNumber (2)-0761
RTECSNumber MO8450000
TSCA No