missing translation for 'onlineSavingsMsg'
Learn More

Vinyl Chloroacetate (stabilized with MEHQ) 99.0+%, TCI America™

Catalog No. C0627500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0627500G 500 g
C062725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0627500G Supplier TCI America Supplier No. C0627500G
Only null left

Chemical Identifiers

CAS 2549-51-1
Molecular Formula C4H5ClO2
Molecular Weight (g/mol) 120.532
MDL Number MFCD00045275
InChI Key XJELOQYISYPGDX-UHFFFAOYSA-N
Synonym vinyl chloroacetate, acetic acid, chloro-, ethenyl ester, acetic acid, 2-chloro-, ethenyl ester, acetic acid, chloro-, vinyl ester, chloroacetic acid vinyl ester, vinyl chloroacetate polymer, vinyl monochloroacetate, chloroacetic acid vinyl, acetic acid, chloro-, vinyl ester, polymers, acmc-1clv7
PubChem CID 82156
IUPAC Name ethenyl 2-chloroacetate
SMILES C=COC(=O)CCl

Specifications

Color Colorless
Boiling Point 136°C
Quantity 500 g
UN Number 2589
Formula Weight 120.53
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Vinyl Chloroacetate (stabilized with MEHQ)
EINECSNumber (2)-3152
RTECSNumber AG2850000
TSCA Yes