missing translation for 'onlineSavingsMsg'
Learn More

Vinyl chloroacetate, 99%, stab. with 4-methoxyphenol

Catalog No. AAL0944618
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0944618 50 g
AAL0944609 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0944618 Supplier Thermo Scientific Chemicals Supplier No. L0944618
Only null left

CAS: 2549-51-1 | C4H5ClO2 | 120.532 g/mol

Vinyl chloroacetate was used to study the effect of the electron-withdrawing groups on the ligand in a series of bis(acetylacetonate)cobalt(II) derivatives. It is used for the enzyme-catalyzed, regioselective acylation of carbohydrates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Vinyl chloroacetate was used to study the effect of the electron-withdrawing groups on the ligand in a series of bis(acetylacetonate)cobalt(II) derivatives. It is used for the enzyme-catalyzed, regioselective acylation of carbohydrates.

Solubility
Difficult to mix in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 2549-51-1
Molecular Formula C4H5ClO2
Molecular Weight (g/mol) 120.532
MDL Number MFCD00045275
InChI Key XJELOQYISYPGDX-UHFFFAOYSA-N
Synonym vinyl chloroacetate, acetic acid, chloro-, ethenyl ester, acetic acid, 2-chloro-, ethenyl ester, acetic acid, chloro-, vinyl ester, chloroacetic acid vinyl ester, vinyl chloroacetate polymer, vinyl monochloroacetate, chloroacetic acid vinyl, acetic acid, chloro-, vinyl ester, polymers, acmc-1clv7
PubChem CID 82156
IUPAC Name ethenyl 2-chloroacetate
SMILES C=COC(=O)CCl

Specifications

Density 1.19
Boiling Point 136°C
Flash Point 51°C (123°F)
Odor Pungent
Refractive Index 1.4435
Quantity 50 g
UN Number UN2589
Beilstein 1741679
Solubility Information Difficult to mix in water.
Formula Weight 120.54
Percent Purity 99%
Chemical Name or Material Vinyl chloroacetate, stabilized with 4-methoxyphenol
Show More Show Less
Hazard Category H226-H302-H315-H317-H318-H335-H341-H351
Hazard Statement GHS H Statement
H300-H310-H330-H314-H318-H226
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P272-P280g-P281-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P310-P330-P333+P313-P363-P370+P37
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: VINYL CHLOROACETATE
EINECSNumber 219-834-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only