missing translation for 'onlineSavingsMsg'
Learn More

Vinyl Benzoate (stabilized with MEHQ) 99.0+%, TCI America™

Catalog No. p-7118815
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
B103225ML 25 mL
B1032500ML 500 mL
2 options

Catalog No. B103225ML

Supplier: TCI America B103225ML

Only null left

Chemical Identifiers

CAS 769-78-8
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.161
MDL Number MFCD00048141
InChI Key KOZCZZVUFDCZGG-UHFFFAOYSA-N
Synonym vinyl benzoate, benzoic acid vinyl ester, benzoic acid, ethenyl ester, benzoic acid, vinyl ester, unii-f1e7c1ggku, vinylester kyseliny benzoove, vinylester kyseliny benzoove czech, f1e7c1ggku, benzoic acid, ethenyl ester 9ci, benzoic acid vinyl
PubChem CID 13037
ChEBI CHEBI:84279
IUPAC Name ethenyl benzoate
SMILES C=COC(=O)C1=CC=CC=C1

Specifications

Melting Point -32°C
Color Colorless
Boiling Point 203°C
Quantity 25 mL
Formula Weight 148.16
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Vinyl Benzoate (stabilized with MEHQ)
EINECSNumber (3)-3793
RTECSNumber DI1050000
TSCA No