missing translation for 'onlineSavingsMsg'
Learn More

Vinyl Benzoate (stabilized with MEHQ) 99.0+%, TCI America™

Catalog No. B103225ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B103225ML 25 mL
B1032500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B103225ML Supplier TCI America Supplier No. B103225ML

Chemical Identifiers

CAS 769-78-8
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.161
MDL Number MFCD00048141
InChI Key KOZCZZVUFDCZGG-UHFFFAOYSA-N
Synonym vinyl benzoate, benzoic acid vinyl ester, benzoic acid, ethenyl ester, benzoic acid, vinyl ester, unii-f1e7c1ggku, vinylester kyseliny benzoove, vinylester kyseliny benzoove czech, f1e7c1ggku, benzoic acid, ethenyl ester 9ci, benzoic acid vinyl
PubChem CID 13037
ChEBI CHEBI:84279
IUPAC Name ethenyl benzoate
SMILES C=COC(=O)C1=CC=CC=C1

Specifications

Melting Point -32°C
Color Colorless
Boiling Point 203°C
Quantity 25 mL
Formula Weight 148.16
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Vinyl Benzoate (stabilized with MEHQ)
EINECSNumber (3)-3793
RTECSNumber DI1050000
TSCA No