missing translation for 'onlineSavingsMsg'
Learn More

Vinyl Acetate Monomer (stabilized with HQ) 99.0+%, TCI America™

Catalog No. A0045500ML
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0045500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A0045500ML Supplier TCI America Supplier No. A0045500ML

Chemical Identifiers

CAS 108-05-4
Molecular Formula C4H6O2
Molecular Weight (g/mol) 86.09
MDL Number MFCD00008713
InChI Key XTXRWKRVRITETP-UHFFFAOYSA-N
Synonym vinyl acetate, acetic acid ethenyl ester, acetic acid vinyl ester, ethenyl ethanoate, 1-acetoxyethylene, vinyl ethanoate, acetoxyethene, vinylacetat, vinyl acetate monomer, vinyl a monomer
PubChem CID 7904
ChEBI CHEBI:46916
IUPAC Name ethenyl acetate
SMILES CC(=O)OC=C

Specifications

Melting Point -93°C
Color Colorless
Boiling Point 72°C
Quantity 500 mL
UN Number 1301
Formula Weight 86.09
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Vinyl Acetate Monomer (stabilized with HQ)
EINECSNumber (2)-0728
RTECSNumber AK0875000
TSCA Yes