missing translation for 'onlineSavingsMsg'
Learn More

Vinpocetine 98.0+%, TCI America™

Catalog No. V0115100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V0115100MG 100 mg
V01151G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V0115100MG Supplier TCI America Supplier No. V0115100MG

Chemical Identifiers

CAS 42971-09-5
Molecular Formula C22H26N2O2
Molecular Weight (g/mol) 350.462
MDL Number MFCD00211233
InChI Key DDNCQMVWWZOMLN-IRLDBZIGSA-N
Synonym vinpocetine, cavinton, ceractin, apovincaminic acid ethyl ester, ethyl apovincamin-22-oate, tcv-3b, ethyl +-apovincaminate, vinpocetinum, bravinton, vinpocetinum inn-latin
PubChem CID 443955
SMILES CCC12CCCN3C1C4=C(CC3)C5=CC=CC=C5N4C(=C2)C(=O)OCC

Specifications

Melting Point 151°C
Color White-Yellow
Quantity 100 mg
Formula Weight 350.46
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Vinpocetine
RTECSNumber JW4792000
TSCA No