missing translation for 'onlineSavingsMsg'
Learn More

Vincristine sulfate, 97+%

Catalog No. p-7041975
Change view
Click to view available options
Quantity:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60907MA 10 mg
AAJ60907MB 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60907MA Supplier Thermo Scientific Chemicals Supplier No. J60907MA

An anti-tumor alkaloid used in cancer research | CAS: 2068-78-2 | C46H58N4O14S | 923.044 g/mol

Vincristine sulfate, is used as an anticancer agent, microtubule disrupter, Induces apoptosis in human lymphoma cells. Other applications include as a cell cycle arresting, apoptotic inducing alkaloid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in DMSO at 60mg/ml. Soluble in water at 25mg/ml with warming. Very poorly soluble in ethanol

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away fom strong oxidizing agents, heavy metal salts.

Chemical Identifiers

CAS 2068-78-2
Molecular Formula C46H58N4O14S
Molecular Weight (g/mol) 923.044
MDL Number MFCD00084729
InChI Key AQTQHPDCURKLKT-DJOZZVTQSA-N
PubChem CID 126963557
SMILES CCC1(CC2CC(C3=C(CCN(C2)C1)C4=CC=CC=C4N3)(C5=C(C=C6C(=C5)C78CCN9C7C(C=CC9)(C(C(C8N6C=O)(C(=O)OC)O)OC(=O)C)CC)OC)C(=O)OC)O.OS(=O)(=O)O

Specifications

Type Vincristine Sulfate
Physical Form Powder
Melting Point >300°C
Color White
Quantity 10 mg
UN Number UN2811
Boiling Point 273°C to 281°C
Beilstein 3924631
Merck Index 14,9986
Solubility Information Soluble in DMSO at 60mg/ml; Soluble in water at 25mg/ml with warming; Very poorly soluble in ethanol
Formula Weight 923.04
Percent Purity ≥98%
Chemical Name or Material Vincristine sulfate
Show More Show Less
Hazard Category H301-H341-H361fd
Hazard Statement GHS H Statement
H360-H341
May damage fertility or the unborn child.
Suspected of causing genetic defects.
Precautionary Statement P201-P202-P264b-P270-P281-P301+P310-P308+P313-P330-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 218-190-0
RTECSNumber OH6340000
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only