missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Vincristine sulfate, 95%

Catalog No. AC203440050
Change view
Click to view available options
Quantity:
5 mg
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC203440050 5 mg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC203440050 Supplier Thermo Scientific Chemicals Supplier No. 203440050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2068-78-2 | C46H56N4O10·H2SO4 | 923.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2068-78-2
Molecular Formula C46H56N4O10·H2SO4
Molecular Weight (g/mol) 923.03
MDL Number MFCD08706469
InChI Key AQTQHPDCURKLKT-DJOZZVTQSA-N
Synonym vincristine sulfate, methyl 9r,10s,11r,12r,19r-11-acetyloxy-12-ethyl-4-13s,15r,17s-17-ethyl-17-hydroxy-13-methoxycarbonyl-1,11-diazatetracyclo 13.3.1.0?, 1 2 .0?, 1 ? nonadeca-4 12 ,5,7,9-tetraen-13-yl-8-formyl-10-hydroxy-5-methoxy-8,16-diazapentacyclo 10.6.1.0 1 ,?.0 2 ,?.0
PubChem CID 126963557
SMILES CCC1(CC2CC(C3=C(CCN(C2)C1)C4=CC=CC=C4N3)(C5=C(C=C6C(=C5)C78CCN9C7C(C=CC9)(C(C(C8N6C=O)(C(=O)OC)O)OC(=O)C)CC)OC)C(=O)OC)O.OS(=O)(=O)O

Specifications

Melting Point >300°C
Color White to Yellow
Boiling Point 273°C to 281°C
Loss on Drying 12% max.
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Quantity 5 mg
Merck Index 15, 10183
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol,slightly soluble in alcohol,practically insoluble in ether
Formula Weight 923.03
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material Vincristine sulfate, 95%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
May damage the unborn child.
Precautionary Statement GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 218-190-

RUO – Research Use Only