missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Vinblastine Sulfate, 98%
Description
Anticancer agent; microtubule disrupter; Induces apoptosis.Vinblastine sulfate is therapeutically used as an antineoplastic. It is also used in the treatment of Hodgkin disease, choriocarcinoma, acute and chronic leukemias.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Quantity | 50 mg |
| Description | White |
| Content And Storage | 2°C to 8°C |
| For Use With (Application) | Anticancer agent; microtubule disrupter; Induces apoptosis |
| Formula Weight | 909.06g/mol |
| Melting Point | 267°C |
| Solubility | Soluble in water or DMSO |
| Product Type | Signal Transduction Reagent |
| Form | Powder |
| Assay | 98% |
RUO – Research Use Only