missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Vinblastine sulfate, 96.0-102.0%

Chemical Identifiers

CAS 143-67-9
Molecular Formula C46H60N4O13S
Molecular Weight (g/mol) 909.06
MDL Number MFCD08706468,MFCD00082457,MFCD08706468
InChI Key KDQAABAKXDWYSZ-JKDPCDLQSA-N
Synonym vinblastine sulfate
PubChem CID 131709225
IUPAC Name methyl (1R,9R,10S,11R,12R,19R)-11-(acetyloxy)-12-ethyl-4-[(13S,15S,17S)-17-ethyl-17-hydroxy-13-(methoxycarbonyl)-1,11-diazatetracyclo[13.3.1.0⁴,¹².0⁵,¹⁰]nonadeca-4(12),5,7,9-tetraen-13-yl]-10-hydroxy-5-methoxy-8-methyl-8,16-diazapentacyclo[10.6.1.0¹,⁹.0²,⁷.0¹⁶,¹⁹]nonadeca-2(7),3,5,13-tetraene-10-carboxylate; sulfuric acid
SMILES OS(O)(=O)=O.CC[C@]1(O)C[C@H]2CN(C1)CCC1=C(NC3=CC=CC=C13)[C@@](C2)(C(=O)OC)C1=CC2=C(C=C1OC)N(C)[C@@H]1[C@]22CCN3CC=C[C@](CC)([C@@H]23)[C@@H](OC(C)=O)[C@]1(O)C(=O)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC203450050
SDS View Documents
Thermo Scientific™
203450050
5mg Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC203450250
SDS View Documents
Thermo Scientific™
203450250
25mg Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

143-67-9
909.06
KDQAABAKXDWYSZ-JKDPCDLQSA-N
131709225
OS(O)(=O)=O.CC[C@]1(O)C[C@H]2CN(C1)CCC1=C(NC3=CC=CC=C13)[C@@](C2)(C(=O)OC)C1=CC2=C(C=C1OC)N(C)[C@@H]1[C@]22CCN3CC=C[C@](CC)([C@@H]23)[C@@H](OC(C)=O)[C@]1(O)C(=O)OC
C46H60N4O13S
MFCD08706468,MFCD00082457,MFCD08706468
vinblastine sulfate
methyl (1R,9R,10S,11R,12R,19R)-11-(acetyloxy)-12-ethyl-4-[(13S,15S,17S)-17-ethyl-17-hydroxy-13-(methoxycarbonyl)-1,11-diazatetracyclo[13.3.1.0⁴,¹².0⁵,¹⁰]nonadeca-4(12),5,7,9-tetraen-13-yl]-10-hydroxy-5-methoxy-8-methyl-8,16-diazapentacyclo[10.6.1.0¹,⁹.0²,⁷.0¹⁶,¹⁹]nonadeca-2(7),3,5,13-tetraene-10-carboxylate; sulfuric acid

Specifications

143-67-9
96.0 to 102% (HPLC)
C46H60N4O13S
5mg
−21.70° (21°C c=2,CH3OH)
vinblastine sulfate
KDQAABAKXDWYSZ-JKDPCDLQSA-N
methyl (1R,9R,10S,11R,12R,19R)-11-(acetyloxy)-12-ethyl-4-[(13S,15S,17S)-17-ethyl-17-hydroxy-13-(methoxycarbonyl)-1,11-diazatetracyclo[13.3.1.0⁴,¹².0⁵,¹⁰]nonadeca-4(12),5,7,9-tetraen-13-yl]-10-hydroxy-5-methoxy-8-methyl-8,16-diazapentacyclo[10.6.1.0¹,⁹.0²,⁷.0¹⁶,¹⁹]nonadeca-2(7),3,5,13-tetraene-10-carboxylate; sulfuric acid
131709225
96.0-102%
Authentic
Glass bottle
MFCD08706468,MFCD00082457,MFCD08706468
15, 10179
-21.7
Solubility in water: insoluble. Other solubilities: soluble in alcohols,acetone,ethyl acetate,,chloroform,practically insoluble in petroleum ether
OS(O)(=O)=O.CC[C@]1(O)C[C@H]2CN(C1)CCC1=C(NC3=CC=CC=C13)[C@@](C2)(C(=O)OC)C1=CC2=C(C=C1OC)N(C)[C@@H]1[C@]22CCN3CC=C[C@](CC)([C@@H]23)[C@@H](OC(C)=O)[C@]1(O)C(=O)OC
909.06
909.05
Vinblastine sulfate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child if i

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Con

GHS Signal Word: Danger

EINECSNumber : 205-606-

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only