missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vileda Professional™ CombiSpeed Pro Mop Frame
Designed for use with all pocket and strip mop systems. Vileda Professional™ CombiSpeed Pro Mop Frame is used for cleaning under tables, radiators etc.
Supplier: Vileda Professional™ 143572
Description
- Slim, angled frame ends and symmetrical double axis-improves easy entry into sponge and pocket mops
- Lightweight, low profile design reduces stress on shoulders and backs
- Low cleaning angle improves cleaning beneath and behind obstacles
- Attaches to Vileda Extendable Handles
- Available in size 40cm
Specifications
| Mop Frame | |
| Blue, Gray | |
| 40 cm | |
| CE Flat Mop (Sterile and non-sterile) |