Learn More
Vikan™ Waterfed Aluminum Handle with Barbed Fitting
Catalog No. 17227156
Change view
Click to view available options
Color:
White
Catalog No. Color
17227156 White
1 options
Catalog No. 17227156 Supplier Vikan™ Supplier No. 29915
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Vikan™ Waterfed Aluminum Handle with Barbed Fitting allows water to travel through it for use in conjunction with a condensation squeegee or waterfed brush system.
  • Comes equipped with a barbed fitting for connecting to a hose with an unfinished end
  • Color-coded
Specifications
Product Type Waterfed Handle
Color-coded Yes
Sterility Non-sterile
Certifications/Compliance FDA Compliant
Color White
Description Thread type Euro Hygienic, Barbed Fitting
Length (English) 61.5 in.
Material Polypropylene
Quantity 1 Ea.
Width (English) 1.25 in.