missing translation for 'onlineSavingsMsg'
Learn More

Vikan™ Ultra Hygiene Bench Squeegee

Catalog No. 19830038
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19830038 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19830038 Supplier Vikan™ Supplier No. 71255

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Features ultra-hygienic design that minimizes areas for water and food soils to collect.

  • Seamless, one-piece construction
  • Soft and flexible rubber blade
  • Removes water or food debris from floors and food contact surfaces

Specifications

Product Type Ultra Hygiene Bench Squeegee
Color White
For Use With (Application) Minimizes areas for water and food soils to collect
Material Polypropylene, Rubber