missing translation for 'onlineSavingsMsg'
Learn More

Vikan™ Standard Broom Handle

The handle with attached item reaches to the chin of the user to prevent occupational back injuries.

$52.90 - $56.70

Specifications

Product Type Standard Broom Handle
Autoclavable Autoclavable
For Use With (Equipment) Brooms, Squeegees and Scrapers
Description Hygienic (Euro) Thread
Color-coded Yes
View More Specs

Products 8
Catalog Number Mfr. No. Color Code Length (English) Price Quantity & Availability  
Catalog Number Mfr. No. Color Code Length (English) Price Quantity & Availability  
19830015
View Documents Promotion Details
Vikan™
29379
Black 60 in.
Each for $56.70
Only null left
 
29635730
View Documents Promotion Details
Vikan™
2937 6
Yellow 60 in.
Each for $56.70
Only null left
 
29635729
View Documents Promotion Details
Vikan™
2937 5
White 60 in.
Each for $56.70
Only null left
 
29635728
View Documents Promotion Details
Vikan™
2937 4
Red 60 in.
Each for $56.70
Only null left
 
29635720
View Documents Promotion Details
Vikan™
2935 6
Yellow 51 in.
Each for $56.70
Only null left
 
19830020
View Documents Promotion Details
Vikan™
29359
Black 51 in.
Each for $52.90
Only null left
 
29635718
View Documents Promotion Details
Vikan™
2935 4
Red 51 in.
Each for $52.90
Only null left
 
29635719
View Documents Promotion Details
Vikan™
2935 5
White 51 in.
Each for $52.90
Only null left
 
Description

Description

  • Made from aluminum and polypropylene
  • Used with brooms, squeegees and scrapers
Specifications

Specifications

Standard Broom Handle
Brooms, Squeegees and Scrapers
Yes
Autoclavable
Hygienic (Euro) Thread
Aluminum, Polypropylene
Videos
SDS
Documents

Documents