missing translation for 'onlineSavingsMsg'
Learn More

Vikan™ Standard Broom Handle

Catalog No. 29635730
Change view
Click to view available options
Color Code:
Black
Red
White
Yellow
Length (English):
51 in.
60 in.
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Code Length (English)
29635730 Yellow 60 in.
19830015 Black 60 in.
29635729 White 60 in.
29635728 Red 60 in.
29635720 Yellow 51 in.
19830020 Black 51 in.
29635718 Red 51 in.
29635719 White 51 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 29635730 Supplier Vikan™ Supplier No. 2937 6

missing translation for 'validMainframeMsg'

The handle with attached item reaches to the chin of the user to prevent occupational back injuries.

  • Made from aluminum and polypropylene
  • Used with brooms, squeegees and scrapers

Specifications

Color Code Yellow
Product Type Standard Broom Handle
Length (English) 60 in.
Autoclavable Autoclavable
For Use With (Equipment) Brooms, Squeegees and Scrapers
Description Hygienic (Euro) Thread
Color-coded Yes
Material Aluminum, Polypropylene