missing translation for 'onlineSavingsMsg'
Learn More

Vikan™ One-Piece Ultra Squeegee

Catalog No. 19830036
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19830036 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19830036 Supplier Vikan™ Supplier No. 71609

missing translation for 'validMainframeMsg'

Features ultra-hygienic design that minimizes areas for water and food soils to collect.

  • seamless, one-piece construction
  • soft and flexible rubber blade
  • Removes water or food debris from floors and food contact surfaces

Specifications

Product Type Single-blade Squeegee
Color Black
For Use With (Application) Minimizes areas for water and food soils to collect
Material Polypropylene, Rubber