Learn More
Vikan™ Medium Overhead Brush
Description
- Medium-bristled Clean-In-Place (CIP) brush
- For cleaning overhead pipes when used with a telescopic handle
- Firm bristles of this brush are ideal in moist environments
- The neck is adjustable to be used a different angles
- Color-coded
Specifications
Specifications
| Product Type | Pipe Brush |
| Color-coded | Yes |
| Handle Material | Stainless Steel |
| Sterility | Non-sterile |
| Color | Blue |
| Length (English) | 20.5 in. |
| Quantity | 1 Ea. |
| Size | Medium |