missing translation for 'onlineSavingsMsg'
Learn More

Vikan™ Liquid Transporting Pail

Catalog No. p-7078536
Change view
Click to view available options
Color:
Black
Red
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19830037 Red
19830030 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19830037 Supplier Vikan™ Supplier No. REM56864
Only null left

For transporting cleaning chemicals and hot or cold ingredients.

  • With drip-free spout
  • Hand-grip raised from base and stainless-steel handle
  • Calibrated for variety of measuring units
  • Flat side prevents spillage
  • Stores using coordinating wall bracket

Specifications

Product Type Transporting Pail
Material Polypropylene
Capacity (English) 3 gal.
Color Red
For Use With (Application) transporting cleaning chemicals, hot or cold ingredients
Temperature (Metric) 121°C