missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vikan™ Lid for 3 Gallon Bucket
Description
- Ideal for preventing spillage of materials
- It has a clip system for securing the lid tightly
- For use with 3 Gallon Bucket
- Color-coded
Specifications
Specifications
| Autoclavable | Not Autoclavable |