missing translation for 'onlineSavingsMsg'
Learn More

Vikan™ Lid for 3 Gallon Bucket

Catalog No. 17227523
Change view
Click to view available options
Color:
Green
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17227523 -
17227526 -
17227529 -
17227524 Green
17227525 -
17227527 -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 17227523 Supplier Vikan™ Supplier No. 56871
Only null left

Vikan™ Lid for 3 Gallon Bucket has a clip system for keeping the lid in place when in use.

  • Ideal for preventing spillage of materials
  • It has a clip system for securing the lid tightly
  • For use with 3 Gallon Bucket
  • Color-coded

Specifications

Autoclavable Not Autoclavable