missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vikan™ Lid for 1.5 Gallon Bucket
Vikan™ Lid for 1.5 Gallon Bucket has a clip system for keeping the lid in place when in use.
Supplier: Vikan™ 56896
Description
- Ideal for preventing spillage of materials
- It has a clip system for securing the lid tightly
- For use with 1.5 Gallon Bucket
- Color-coded
Chemical Identifiers
| Not Autoclavable |
Specifications
| Not Autoclavable |