missing translation for 'onlineSavingsMsg'
Learn More

Vikan™ Lid for 1.5 Gallon Bucket

Catalog No. 17227544
Change view
Click to view available options
Color:
Green
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17227544 -
17227547 -
17227546 -
17227542 Green
17227543 -
17227548 -
17227541 -
17227545 -
17227549 -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 17227544 Supplier Vikan™ Supplier No. REM56894

Vikan™ Lid for 1.5 Gallon Bucket has a clip system for keeping the lid in place when in use.

  • Ideal for preventing spillage of materials
  • It has a clip system for securing the lid tightly
  • For use with 1.5 Gallon Bucket
  • Color-coded

Specifications

Autoclavable Not Autoclavable