missing translation for 'onlineSavingsMsg'
Learn More

Vikan™ Lid for 1.5 Gallon Bucket

Vikan™ Lid for 1.5 Gallon Bucket has a clip system for keeping the lid in place when in use.

Supplier:  Vikan™ REM56893

 View more versions of this product

Catalog No. 17227543


Only null left

Description

Description

  • Ideal for preventing spillage of materials
  • It has a clip system for securing the lid tightly
  • For use with 1.5 Gallon Bucket
  • Color-coded
Specifications

Chemical Identifiers

Not Autoclavable

Specifications

Not Autoclavable
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions