Learn More
Vikan™ Ergonomic Shovel with Small Blade
Catalog No. 29635517
Change view
Click to view available options
Color Code:
Blue
Green
Red
White
Yellow
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Code
29635517 White
17227496 Red
17227498 Yellow
17227494 Green
17227495 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 29635517 Supplier Vikan™ Supplier No. 56115
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Vikan™ Ergonomic Shovel with Small Blade is designed without any sharp corners or deep recessess.
  • Designed for use in direct contact with food
  • The handle is reinforced with an inner aluminum tube
  • It is stepped in two places to help prevent back injuries
  • The neck joint has been strengthened in order to withstand heavy-duty use
  • The long and small blade combination makes it suitable for shoveling smaller volumes and reaching into deep containers
  • Color-coded
Specifications
Product Type Ergonomic Shovel
Blade Material Polypropylene
Color Code White
Color-coded Yes
Handle Material Polypropylene, Aluminum
Head Style Square
Length (English) 50 in.
Certifications/Compliance FDA Compliant
No. per Pack 1 Ea.
Weight (English) 3.39 lb.