Learn More
Vikan™ Blade Cleaning Brush with Hand Guard
Vikan™ Blade Cleaning Brush with Hand Guard makes cleaning machinery with dangerous blades safer.
Supplier: Vikan™ 41845
Description
- Ideal for cleaning tools with exposed blades that could cause injury to the user
- The narrow single row of bristles fits perfectly between the blades of any bowl cutter
- The hand guard protects the user's hand from being cut during the cleaning process
- This brush allows the user to clean the machine without disassembly
- A durable polypropylene shield protects hands while the medium bristles thoroughly clean blades
- Color-coded
Specifications
| Cleaning Brush | |
| Yes | |
| Non-sterile | |
| White | |
| 1 Ea. |
| Polyester | |
| Polypropylene | |
| FDA Compliant | |
| 18.9 in. | |
| Medium |