missing translation for 'onlineSavingsMsg'
Learn More

Vikan™ Telescopic Aluminum Handle, 62 to 113 in.

Catalog No. 29635745
Change view
Click to view available options
Color Code:
Black
Blue
Red
Yellow
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Code
29635745 Red
17227143 Yellow
17227140 Blue
17227144 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 29635745 Supplier Vikan™ Supplier No. 2975 4

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Telescopic aluminum handle is a durable and safe option to use in hygiene sensitive areas.

  • For use with pad holder and deck scrubs
  • Suitable for reaching high walls and tanks
  • Cna be used with clean-in-place brushes for cleaning on top of overhead pipes
  • The handle extends to a maximum length of 115 in.
  • Uniquely suited for color-coding and HACCP programs

Specifications

Color Code Red
Product Type Handle
Sterility Non-sterile
Quantity 1 Ea.
Description Thread type Euro Hygienic
Certifications/Compliance FDA Compliant
Color-coded Yes
Material Aluminum, Polypropylene