Learn More
Vikan™ Single Blade Ultra Hygiene Bench Squeegee, 10 in.
Squeegee is ideal for removing water or food debris from food contact surfaces.
Supplier:  Vikan™ 71253
 View more versions of this product
Catalog No. 17227827
Only null left
Description
Description
  • One-piece construction
  • Single-blade squeegee
  • Ultra-hygienic design that minimizes areas for water and food soils to collect
  • Effective in areas requiring the highest level of hygiene
  • The soft and flexible rubber blade is ideal for removing water or food debris from floors and food contact surfaces
  • Hygiene squeegees are a good foundation for any HACCP or food safety program
  • A splash guard helps prevent water from reaching the dried surface
  • Color-coded
Specifications
Chemical Identifiers
Ultra Hygiene Bench Squeegee
Thread type Euro Hygienic
Polypropylene, Rubber
Medium
FDA Compliant
Thread type Euro Hygienic
1 Ea.
3.5 in.
Specifications
Ultra Hygiene Bench Squeegee
Blue
Thread type Euro Hygienic
1 Ea.
3.5 in.
FDA Compliant
Thread type Euro Hygienic
Polypropylene, Rubber
Medium
Product Suggestions
Product Suggestions