missing translation for 'onlineSavingsMsg'
Learn More

Vikan™ Foam Blade Squeegee, 10 in.

Catalog No. 17227898
Change view
Click to view available options
Color:
Blue
Green
Red
White
Yellow
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17227898 Red
17227899 Yellow
17227896 Green
17227897 Blue
19830221 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17227898 Supplier Vikan™ Supplier No. 77514

Squeegee is ideal for drying surfaces after a sanitizing process or for pushing food from one area to another.

  • Provides effective removal of both water and food debris from food preparation surfaces
  • Blade cartridge can be detached for easy cleaning or replacement
  • Replacement squeegee blade
  • Color-coded

Specifications

Product Type Ultra Hygiene Bench Squeegee
Certifications/Compliance FDA Compliant
Color Red
Description Thread type Euro Hygienic
Includes Thread type Euro Hygienic
Material Polypropylene, Foam Rubber
Quantity 1 Ea.
Size Small
Width (English) 3.1 in.