missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ ViiA™ 7 System Automation Accessory Robot (100 to 240V)

Catalog No. 4453551
Encompass
Catalog No. 4453551 Supplier Applied Biosystems™ Supplier No. 4453551

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use with the ViiA™ 7 Real-Time PCR System.

Specifications

Content And Storage Includes Robot, Plate Stacks, and Service Installation.
For Use With (Equipment) ViiA™ 7 System
Product Type Automation Robot
For Use With (Application) Real time PCR (qPCR)
Quantity 1 Ea.