Learn More
ViewRNA™ ISH Cell Assay Kit
Supplier: Invitrogen QVC0001
Specifications
| Components sufficient for 1-plex to 3-plex (AlexaFluor 488, 546 and 647) assay in 24- well plate format (24 samples), 96 assays when using cover slips mounted on glass slides |
|
| 1 kit | |
| Detection Assay Kit |
For Research Use Only. Not for use in diagnostic procedures.