missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Videodrop

Catalog No. 01257546
Change view
Click to view available options
Camera:
Digital Color
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Camera
01257546 Digital Color
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01257546 Supplier Corning™ Supplier No. VD1000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Through the utilization of interferometry in conjunction with microscopy, the Corning™ Microdrop; senses, detects, characterizes, and tracks single nanoparticles in real time.

The Corning™ Videodrop utilizes the principles of interferometry in conjunction with microscopy to sense, detect, characterize, and track single nanoparticles in real time. The Videodrop aligns with the cell and gene therapy segment of the bioproduction market due to the correlation of collecting physical titer of viral vectors such as Lentivirus, Retrovirus, Adenovirus, and Extracellular Vesicles in the range of 80-500 nanometers. Videodrop outputs are linear compared to p24 ELISA, RTqPCR, and NTA assays, as concluded on the application notes within the Resources tab.

Features:

  • Efficient
    • Under 1 minute loading, processing, and cleaning
    • No calibration nor wash needed
  • Repeatable and Reproducible Results
    • No inter-operator bias
    • No parameter adjustment

Specifications

Type BioImaging System
Shaking Linear, Orbital, Double Orbital
Camera Digital Color
CO2 Concentration Range 80 to 500 nm
Color Silver
Wavelength 450 nm
Data Management Several Export Options
Display 11.82 x 11.82 x 31.15 in.
Software Compatibility Windows 10 or 11, 16GB of RAM, Hard drive of 4+ TB, Processor of 8+ cores, USB ss port
Height (English) 31.15 in.
Height (Metric) 79.1 cm
Weight (English) 39.69 lbs
Weight (Metric) 18 kg
Width (English) 11.82 in.
Width (Metric) 30 cm
Certifications/Compliance GMP Facility Requirements
Content And Storage 30 to 85% relative humidity
Warranty Limited
Electrical Requirements 12 V
Voltage 120/230 V
Wattage 4.5 W
Show More Show Less