Learn More
Corning™ Videodrop Glass Slides Box, 50/Cs
Catalog No. 01257549
Change view
Click to view available options
Corning™ Videodrop Glass Slides
Catalog No.
01257549 Corning™ Videodrop Glass Slides
1 options
Catalog No. 01257549 Supplier Corning™ Supplier No. VD1000GS
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specialized glass slide for the Corning™ Videodrop. Only 5 to 10 μL of bioproduction sample is required.
Specialized glass slide for the Corning™ Videodrop. Only 5 to 10 μL of bioproduction sample is required. The notch on the top surface is engineered to maintain the sample drop during analysis and is intended to come in contact with the objective lens.
Specifications
Frosted Not Frosted
Well Count 1
Precleaned Yes
Sterility Non-sterile
Material Glass
Type Microscope Slide
Color Clear