Learn More
Beckman Coulter VI-CELL MetaFLEX Sensor Cassette 300/30
Catalog No. NC1565716
Encompass
Vi-CELL MetaFLEX sensor cassette, SC300 measures pH, pO2, pCO2, Glucose, Lactate, Electrolytes and more parameters associated with whole blood analysis
Catalog No. NC1565716 Supplier Beckman Coulter Supplier No. B93030
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Vi-CELL MetaFLEX sensor cassette, SC300 measures pH, pO2, pCO2, Glucose, Lactate, Electrolytes and more parameters associated with whole blood analysis