Learn More
Microsolv Technology VIALS SCREW TOP AMBER 1.5ML 8- DFS Item
Supplier:  Microsolv Technology Corporation 9582S0AV
Vials, autosampler, amber borosilicate glass, screw top with 8-425mm threads, 2ml 12x32mm. Basik brand. 100/pack.
Catalog No. 5001452404
Only null left
Explore available promotions