Learn More
Microsolv Technology Corporation Vial Decapper 20Mm
Catalog No. 5001451891
Encompass
Decapper Tool, for 20mm Aluminum Headspace Caps. MicroSolv Brand. 1/EA. .
Catalog No. 5001451891 Supplier Microsolv Technology Corporation Supplier No. 95025DC
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Decapper Tool, for 20mm Aluminum Headspace Caps. MicroSolv Brand. 1/EA. .