missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ Vial Crimp Seals, BiMetal (Magnetic)
The sealing surface is aluminum, and the inner diameter is made of tin to add the magnetic surface. Available in a variety of colors.
Supplier: MilliporeSigma™ Supelco™ 29176U
Chemical Identifiers
| 20 mm | |
| 100/Pk. | |
| 20 mm |
| PTFE, Silicone | |
| 3 mm | |
| Crimp Seal |
Specifications
| Blue | |
| PTFE, Silicone | |
| 3 mm | |
| Crimp Seal |
| 20 mm | |
| 100/Pk. | |
| 20 mm |