Learn More
Ab Sciex Llc Insertion Lever Interface Parts Kit
Supplier: Danaher Corp A95348
The vial cap opener assembly contains two vial cap openers and syringes allowing precision alignment and pressure sealing between vial caps and the separation interface. This component also provides protection for both capillary and electrode during instrument use. This vial cap opener is for use only with CESI 8000 PA 800 Enhanced or PA 800 plus instruments.