Learn More
Danaher Corp VIAL CAP BUFFER BLUE
Catalog No. NC9816690
Encompass
Universal vial caps are used in combination with Universal Vials (Part No. A62251). Universal vial caps are designed for use only with PA 800 Plus or PA 800 Enhanced instruments and should not be reused. Package of 100.
Catalog No. NC9816690 Supplier Danaher Corp Supplier No. A62250
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Universal vial caps are used in combination with Universal Vials (Part No. A62251). Universal vial caps are designed for use only with PA 800 Plus or PA 800 Enhanced instruments and should not be reused. Package of 100.