Learn More
Tradewinds Direct Inc BioX Tricode 0.5mL Cryovial, Yellow Cap, External Thread. Case of 1000
Catalog No. 502274237
Encompass
BioX 0.5mL clear polypropylene sterile pre-set 1D & 2D barcoded cryovials with external thread and yellow caps assembled. Cryovials have writing patch and marked graduations. 25 cryovials/bag, 20 bags/pack, 2 packs/case, price/case
Catalog No. 502274237 Supplier Tradewinds Direct Inc Supplier No. BX889054
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BioX 0.5mL clear polypropylene sterile pre-set 1D & 2D barcoded cryovials with external thread and yellow caps assembled. Cryovials have writing patch and marked graduations. 25 cryovials/bag, 20 bags/pack, 2 packs/case, price/case