Learn More
Biotium ViaFluor 647, 50 uL
Catalog No. NC1663132
Encompass
ViaFluor Live Cell Microtubule Stains are cell-permeable taxol probes for imaging the microtubule cytoskeleton in live cells. Available with blue, green, or far-red fluorescence.
Catalog No. NC1663132 Supplier Biotium Supplier No. 70063T
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
ViaFluor Live Cell Microtubule Stains are cell-permeable taxol probes for imaging the microtubule cytoskeleton in live cells. Available with blue, green, or far-red fluorescence.