Learn More
Beckman Coulter Canada Lp Vi-CELL XR Single Reagent Pack with 120 Sample Vials DFS Item
Supplier:  Beckman Coulter 383260
Beckman Coulter Vi-CELL Reagent Single Pak. This reagent pack does not include any sample cups. The reagent kit can be used on any Beckman Coulter automated cell viability analyzer, ViCELL, including Vi-CELL S, Vi-CELL AS, and Vi-CELL XR models. The Beckman Vi-CELL is a completely automated cell counter that utilizers the typan blue method for fast, accurate results.
Catalog No. NC9263868
Only null left
Explore available promotions