missing translation for 'onlineSavingsMsg'
Learn More

3M™ VFlex™ 1804 Series N95 Healthcare Particulate Respirators and Surgical Masks

Catalog No. 19040629
Change view
Click to view available options
Size:
Standard
Small
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19040629 Small
19040625 Standard
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19040629 Supplier 3M™ Supplier No. 7100145154
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

N95 surgical masks provide respiratory protection against certain airborne particles. The folded style expands for a spacious feel and the pleats help increase surface area for ease of breathing.

  • NIOSH approved N95, small or standard size
  • FDA cleared for use as surgical mask
  • Fluid resistant and disposable
  • 3M proprietary filter media and pleats help make breathing easier
  • Uniquely designed tabs for respirator positioning on the face
  • Pleats flex with mouth movement while talking
  • Flatfold design allows for convenient storage prior to use

Specifications

Aerosol Type N - Not Resistant to Oil
Assigned Protection Factor (APF) 10
Certifications/Compliance ASTM F1862
Certifications (CSA) Health Canada Class I Medical Device
Certifications (NIOSH) NIOSH 42 CFR 84, N95, approval number: TC-84A-7789
Certifications (OSHA) OSHA 1910.134
Color White
Design Type Flat-fold
Disposable Single-use
Filter Type Surgical N95
Material Polypropylene
Product Type Healthcare Particulate Respirator and Surgical Mask
Size Small
With Exhalation Valve No
Filter Efficiency 95%
For Use With (Application) Grinding, sanding, sweeping, bagging and other dusty or arid applications
Latex Free Yes
Material Headstrap Polyisoprene
Model 1804S
Nose Piece Material Aluminum
Nosepiece Type Adjustable Nose Clip
Quantity 400/Cs.
Resistant To Fluid 80 mmHg
Size (English) 3.75 tall x 5.75 wide x 2 in. deep (approx.)
Sterility Non-sterile
Strap Attachment Type Stapled Headbands
FDA Cleared Surgical Mask Yes
Show More Show Less