missing translation for 'onlineSavingsMsg'
Learn More

Bulwark™ FR Men's Nomex™ IIIA Uniform Shirt, Royal Blue

Catalog No. 191661058
Change view
Click to view available options
Closure Type:
Button
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Type
191661058 Button
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 191661058 Supplier Bulwark™ FR Supplier No. SND6RB RG S

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Enjoy this classic Uniform Shirt made to withstand your work while providing CAT 1 AR/FR, NFPA™ 2112. It features a banded, topstitched collar as well as two chest pockets with button flap closures and sewn in pencil stall.

  • Certifications Level 1: NFPA 2112, NFPA 70E
  • Certifications Level 2: Category 1 Protection
  • Hazard:
  • Primary Closure: Button
  • Wash Care: Industrial Laundry - Light Soil, Home Wash
  • Fabric: Flame-resistant, 6 oz. (205 g/m2) Nomex™ IIIA
  • Blend: 93% Nomex™ Aramid/5% Kevlar™ Aramid/2% Other Fiber
  • Finish: Autoclave
  • Closure: Placket front with button closure as well as topstitched cuffs with adjustable button closures
  • Collar: Banded, topstitched collar
  • Features:
    • Banded, topstitched collar
    • Topstitched cuffs with adjustable button closures
    • Placket front with button closure
    • Tailored sleeve placket
    • Autoclaved
  • Product Collection: Nomex™
  • Protection: ATPV Arc Rating: 5.2
  • Country of Origin: Imported, Made in USA Fabric
  • Fit Details: Regular Fit
  • Gender: Male
  • Materials: Nomex™
  • Brand: Bulwark™

Specifications

Size Small
Gender Men's
Material Nomex
Color Royal Blue
Neck Style Collar
Sleeve Style Long Sleeve
Closure Type Button
No. of Pockets 2
Resistance Flame
Certifications/Compliance NFPA 2112, NFPA 70E, Category 1 Protection
Style Regular
Show More Show Less