missing translation for 'onlineSavingsMsg'
Learn More

VETRANAL™ Triflupromazine Hydrochloride Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. p-7214112
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022534 10 mg
111022535 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111022534 Supplier Merck Emd Millipore Supplier No. 4697610MG
Only null left

Specifications

CAS 1098-60-8
Molecular Formula C18H19F3N2S · HCl
Linear Formula C18H19F3N2S · HCl
MDL Number MFCD00058103
Quantity 10 mg
UN Number UN 2811 6.1/PGIII
Synonym N,N-Dimethyl-2-(trifluoromethyl)-10 H-phenothiazine-10-propanamine monohydrochloride
Formula Weight 388.88
Grade Analytical Standard
Physical Form Neat

VETRANAL is a registered trademark of Sigma-Aldrich Chemie GmbH