missing translation for 'onlineSavingsMsg'
Learn More

VETRANAL™ Toltrazuril sulfoxide, MilliporeSigma™ Supelco™

Catalog No. 111015188
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015188 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015188 Supplier Merck Emd Millipore Supplier No. 3381510MG
Only null left

Toltrazuril sulfoxide is a metabolite of the antiparasitic veterinary drug, toltrazuril.

Pharmaceutical (small molecule)

Specifications

Product Line VETRANAL
CAS 69004-15-5
Physical Form Neat
Linear Formula C18H14F3N3O5 S
MDL Number MFCD10567359
Synonym 1-Methyl-3-{3-methyl-4-[4-(trifluormethylsulfinyl)-phenoxy]-phenyl}-1,3,5-triazin-2,4,6(1 H,3 H,5 H)-trion
Type Analytical Standard
Quantity 10 mg
Formula Weight 441.38
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH