missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VETRANAL™ Tinidazole, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| Product Line | VETRANAL |
| CAS | 19387-91-8 |
| Physical Form | Neat |
| Linear Formula | C8H13N3O4 S |
| MDL Number | MFCD00057217 |
| Synonym | 1-[2-(Ethylsulfonyl)ethyl]-2-methyl-5-nitroimidazole |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 247.27 |
| Grade | Analytical Standard |
VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH