missing translation for 'onlineSavingsMsg'
Learn More

VETRANAL™ Sulfadoxin, MilliporeSigma™ Supelco™

Catalog No. 111013679
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013679 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013679 Supplier Merck Emd Millipore Supplier No. 31736250MG
Only null left

Sulfadoxin belongs to the long acting group of sulfonamides, which acts by inhibiting dihydropteroate synthase in intracellular sporozoa such as Eimeria and Toxoplasma and Plasmodium.

Pharmaceutical (small molecule)

Specifications

Product Line VETRANAL
CAS 2447-57-6
Physical Form Neat
Linear Formula C12H14N4O4 S
MDL Number MFCD00792890
Synonym 4-Amino-N-(5,6-dimethoxy-4-pyrimidinyl)benzenesulfonamide; Sulfadoxine
Type Analytical Standard
Quantity 250 mg
Formula Weight 310.33
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH