missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VETRANAL™ Nequinate, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| Product Line | VETRANAL |
| CAS | 13997-19-8 |
| Physical Form | Neat |
| Linear Formula | C22H23 NO4 |
| MDL Number | MFCD00867198 |
| Synonym | Methyl 7-(benzyloxy)-6-butyl-1,4-dihydro-4-oxo-3-quinolinecarboxylate; Methyl benzoquate |
| Type | Analytical Standard |
| Quantity | 25 mg |
| Formula Weight | 365.42 |
| Grade | Analytical Standard |
VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH